![(2R,2′R)-2,2′-Bipyrrolidine ≥99.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/291/192/6febe0bc-3608-44ae-888f-78c8398bf82e/640/6febe0bc-3608-44ae-888f-78c8398bf82e.png)
(2R,2′R)-2,2′-Bipyrrolidine
![(2R,2′R)-2,2′-Bipyrrolidine ≥99.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/291/192/6febe0bc-3608-44ae-888f-78c8398bf82e/640/6febe0bc-3608-44ae-888f-78c8398bf82e.png)
Synonym(s):
(R,R)-2,2′-Bipyrrolidine, (R,R)-2,2′-Bipyrrolidinyl
Empirical Formula (Hill Notation):
C8H16N2
CAS Number:
Molecular Weight:
140.23
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing