![2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic acid pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/303/197/4983f71e-abfb-4dc7-b1e9-0446b7f2c476/640/4983f71e-abfb-4dc7-b1e9-0446b7f2c476.png)
2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic acid
![2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic acid pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/303/197/4983f71e-abfb-4dc7-b1e9-0446b7f2c476/640/4983f71e-abfb-4dc7-b1e9-0446b7f2c476.png)
Synonym(s):
2-[(2-Bromo-6-chlorophenyl)amino]benzeneacetic acid
Empirical Formula (Hill Notation):
C14H11BrClNO2
CAS Number:
Molecular Weight:
340.60
Products
Brand
Formula Weight
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing