Skip to Content
Merck
1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one

1-Ethyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C15H18FN3O
CAS Number:
Molecular Weight:
275.32

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade