Skip to Content
Merck
1,4-Dichlorobenzene-d4 98 atom % D, 98% (CP)

1,4-Dichlorobenzene-d4

1,4-Dichlorobenzene-d4 98 atom % D, 98% (CP)
Synonym(s):
Tetradeutero-1,4-dichlorobenzene
Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing