Product Name
Diethofencarb PESTANAL®, analytical standard
Product Number
34087
Product Brand
sial
CAS Number
87130-20-9
Molecular Weight
267.32
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT97 - 103 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS