Product Name
1-Benzyl-3-hydroxy-1H-indazole 97%
Product Number
722219
Product Brand
aldrich
CAS Number
2215-63-6
Molecular Weight
224.26
Test | Specification |
---|---|
Appearance (Color) | White to Off-White to Beige |
Appearance (Form) | Powder to Crystalline Powder |
Infrared spectrum | Conforms to Structure |
Carbon | 72.4 - 77.6 % |
Nitrogen | 12.0 - 13.0 % |
GC (area %) | ≥96.5 % |