Product Name
2-Bromo-3-pyridyl trifluoromethanesulfonate 96%
Product Number
649945
Product Brand
aldrich
CAS Number
157373-97-2
Molecular Weight
306.06
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 95.5-104.5% (WITH AGNO3 AFTER OXYGEN |
COMBUSTION) | |
HIGH PRESSURE LIQUID | 95.5% (MINIMUM) |
APPROVED OCTOBER 31, 2006 DJP |