SAB5702601
Anti-CAD Antibody, clone 0O8D6, Rabbit Monoclonal
Synonym(s):
CDG1Z, EIEE50, GATD4
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
0O8D6, monoclonal
form
liquid
mol wt
250 kDa
species reactivity
mouse, mouse
concentration
1.50 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
MAALVLEDGSVLRGQPFGAAVSTAGEVVFQTGMVGYPEALTDPSYKAQILVLTYPLIGNYGIPPDEMDEFGLCKWFESSGIHVAALVVGECCPTPSHWSA
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CAD(790)
General description
Immunogen
Application
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service