ENA341295462
Enamine
5-[(3-bromophenoxy)methyl]furan-2-carbaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-[(3-bromophenoxy)methyl]-2-furaldehyde
Inchi Code
1S/C12H9BrO3/c13-9-2-1-3-10(6-9)15-8-12-5-4-11(7-14)16-12/h1-7H,8H2
InChI key
JBRYFXVNQUHENM-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-92250
MSDS
https://www.enaminestore.com/catalog/EN300-92250
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service