Merck
HomeProfessor Product PortalEllman Group – Professor Product Portal
Ellman Group – Professor Product Portal
Jonathan Ellman
Professor Jonathan A. Ellman
The Ellman group has participated in the development of a variety of C-H functionalization methods.  An electron rich phosphine ligand has proven to be very useful for a variety of Rh(I)-catalyzed C-C bond forming reactions applicable to heterocycle synthesis as exemplified in the recent Science paper “Proton Donor Acidity Controls Selectivity in Nonaromatic Nitrogen Heterocycle Synthesis.” Another useful ligand developed for the highly functional group compatible direct arylation of nitrogen heterocycles is described in a 2008 J. Am. Chem. Soc.  paper “Rh(I)-Catalyzed Arylation of Heterocycles via C-H Bond Activation: Expanded Scope through Mechanistic Insight.”
The Ellman group also developed the chiral amine reagent tert-Butanesulfinamide, which is extensively used in academics and industry for the asymmetric synthesis of amines. A comprehensive survey of tert-Butanesulfinamide methods and applications up through 2009 is provided in the 2010 Chemical Reviews article, “Synthesis and Applications of tert-Butanesulfinamide.”
Ellman Group Website
Recent papers from the Ellman Group
1.
Duttwyler S, Chen S, Takase MK, Wiberg KB, Bergman RG, Ellman JA. 2013. Proton Donor Acidity Controls Selectivity in Nonaromatic Nitrogen Heterocycle Synthesis. Science. 339(6120):678-682. https://doi.org/10.1126/science.1230704
2.
Robak MT, Herbage MA, Ellman JA. 2010. Synthesis and Applications of tert-Butanesulfinamide. Chem. Rev.. 110(6):3600-3740. https://doi.org/10.1021/cr900382t
Products available from the Ellman Group:
Sorry, an unexpected error has occurred
Network error: Failed to fetch
DISCIPLINES
  • Organic Synthetic Chemistry
Professor Product Portal Index