Merck
UHPLC Analysis of Fat Soluble Vitamins on Titan C18, Acetonitrile Mobile Phase
UHPLC Analysis of Fat Soluble Vitamins on Titan C18, Acetonitrile Mobile Phase application for UHPLC
Materials
standard
Product No.
Description
Pricing
DL-α-Tocopherol acetate
≥96% (HPLC)
Retinyl acetate
analytical standard
(±)-α-Tocopherol
analytical standard
(+)-γ-Tocopherol
analytical standard
δ-Tocopherol
analytical standard
CONDITIONS
column
TITAN C18, 50 mm x 2.1mm I.D. x 1.9 μm particles (577122-U)
mobile phase
100% acetonitrile
flow rate
0.4 mL/min
pressure
1755 psi (121 bar)
column temp.
30 °C
detector
UV, 280 nm
injection
0.5 μL
sample
All compounds at 0.5 mg/mL except retinyl acetate at 0.05 mg/mL in methanol
Description
Legal Information
null