Merck
HPLC Analysis of Adenosine Nucleosides on Supelcosil LC-18-T
HPLC Analysis of Adenosine Nucleosides on Supelcosil™ LC-18-T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-18-T HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
2′-Deoxyadenosine 5′-monophosphate
Sigma Grade, 98-100%
Adenosine 5′-triphosphate disodium salt hydrate
BioXtra, ≥99% (HPLC), from microbial
Adenosine 5′-diphosphate sodium salt
bacterial, ≥95% (HPLC)
CONDITIONS
column
SUPELCOSIL LC-18-T, 15 cm x 4.6 mm I.D, 3 μm particles (58970-U)
mobile phase
[A] 0.1 M potassium phosphate, pH 7; [B] acetonitrile
gradient
0 to 2.5% B in 4 min, to 7.5% B in 2 min, to 0% B in 0.1 min, held at 0% B for 3.9 min
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
20 μL
sample
each compound 5 μg/mL in water
Description
Analysis Note
A need to analyze nucleosides separated from plant and animal products for pediatric nutritional and other research interests exists. SUPELCOSIL LC-18-T columns have proven effective in the separation of adenosine nucleosides.
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC