Merck
HPLC Analysis of Vitamins, Fat Soluble, on Ascentis® Si
HPLC Analysis of Vitamins, Fat Soluble, on Ascentis® Si application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
(+)-γ-Tocopherol
≥96% (HPLC)
α-Tocopherol
≥95.5%
Cholecalciferol (D3)
analytical standard
Menadione (K3)
analytical standard
CONDITIONS
column
Ascentis Si, 15 cm x 4.6 mm I.D., 5 μm particles (581512-U)
mobile phase
[A] hexane; [B] ethyl acetate
gradient
10 to 30% B in 10 min; held for 2 min; to 10% B in 1 min
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 290 nm
injection
10 μL
sample
as indicated in 96:4 (hexane:isopropanol)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany