Merck
HPLC Analysis of Coumachlor Enantiomers on Astec® CYCLOBOND I 2000 DM
HPLC Analysis of Coumachlor Enantiomers on Astec® CYCLOBOND I 2000 DM application for HPLC
Materials
standard
Product No.
Description
Pricing
Coumachlor
98%
CONDITIONS
column
CYCLOBOND I 2000 DM, 25 cm x 4.6 mm I.D., 5 μm particles (20924AST)
mobile phase
[A] acetonitrile; [B] 20 mM ammonium acetate, pH 2.9; (20:80, A:B)
flow rate
0.8 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
3 μL
sample
coumachlor, 1 mg/mL in acetonitrile:water (50:50)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null