Merck
GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm)
GC Analysis of Closely Eluting Analytes from US EPA Method 8270 on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm) suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
SLB-5ms, 15 m x 0.10 mm I.D., 0.10 μm (28466-U)
oven
40 °C (2 min), 22 °C/min to 240 °C, 10 °C/min to 330 °C (1 min)
inj. temp.
250 °C
detector
FID, 330 °C
carrier gas
helium, 30 cm/sec @ 200 °C, set using methane
injection
0.1 μL splitless (0.75 min)
liner
2 mm I.D., straight
sample
50 ng on-column of a 72 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column), in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany