- Identification of a novel allele HLA-B*7805 in a Japanese female.
Identification of a novel allele HLA-B*7805 in a Japanese female.
Tissue antigens (2002-02-15)
T Sekimoto, M Araseki, S Sato, S Sakaya, T Kato, H Ikeda
PMID11844148
ABSTRACT
A new HLA-B*78 allele, B*7805, was identified in a healthy Japanese female. The results of her serological HLA class I typing showed an unusual Bw4/Bw6 pattern with strongly positive reactivity to anti-Bw6, i.e. A24, -, B52, -, Bw4, Bw6. In DNA typing, she was typed as A*24, -, B*52, B*78-like, Cw1202, -, (Bw4, Bw6). Cloning and sequencing of exon 2 and exon 3 of her B locus genes revealed a new allele B*7805. The cloned B*7805 differed from B*78021 by three nucleotide substitutions in exon 2 at position 259 (A to G), 261 (C to G) and 272 (A to C), and contained sequences defining Bw6 motif in the region of codon 77 to 83.