Merck
  • The unfolded protein response impacts melanoma progression by enhancing FGF expression and can be antagonized by a chemical chaperone.
The unfolded protein response impacts melanoma progression by enhancing FGF expression and can be antagonized by a chemical chaperone.
Scientific reports (2017-12-14)
Karin Eigner, Yüksel Filik, Florian Mark, Birgit Schütz, Günter Klambauer, Richard Moriggl, Markus Hengstschläger, Herbert Stangl, Mario Mikula, Clemens Röhrl
ABSTRACT
The mechanisms hallmarking melanoma progression are insufficiently understood. Here we studied the impact of the unfolded protein response (UPR) - a signalling cascade playing ambiguous roles in carcinogenesis - in melanoma malignancy. We identified isogenic patient-derived melanoma cell lines harboring BRAF
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
4-Phenylbutyric acid, 99%