Merck
  • The first wave of T lymphopoiesis in zebrafish arises from aorta endothelium independent of hematopoietic stem cells.
The first wave of T lymphopoiesis in zebrafish arises from aorta endothelium independent of hematopoietic stem cells.
The Journal of experimental medicine (2017-09-22)
Ye Tian, Jin Xu, Shachuan Feng, Sicong He, Shizheng Zhao, Lu Zhu, Wan Jin, Yimei Dai, Lingfei Luo, Jianan Y Qu, Zilong Wen
ABSTRACT
T lymphocytes are key cellular components of the adaptive immune system and play a central role in cell-mediated immunity in vertebrates. Despite their heterogeneities, it is believed that all different types of T lymphocytes are generated exclusively via the differentiation of hematopoietic stem cells (HSCs). Using temporal-spatial resolved fate-mapping analysis and time-lapse imaging, here we show that the ventral endothelium in the zebrafish aorta-gonad-mesonephros and posterior blood island, the hematopoietic tissues previously known to generate HSCs and erythromyeloid progenitors, respectively, gives rise to a transient wave of T lymphopoiesis independent of HSCs. This HSC-independent T lymphopoiesis occurs early and generates predominantly CD4 T
Sigma-Aldrich
N,N′-Bis(3-pentyl)perylene-3,4,9,10-bis(dicarboximide), 98% (HPLC)