Merck
  • Methylxanthines induce structural and functional alterations of the cardiac system in zebrafish embryos.
Methylxanthines induce structural and functional alterations of the cardiac system in zebrafish embryos.
BMC pharmacology & toxicology (2017-11-17)
Ram Manohar Basnet, Daniela Zizioli, Michela Guarienti, Dario Finazzi, Maurizio Memo
ABSTRACT
Zebrafish embryos are emerging as a model for pharmacological and toxicological studies. We used zebrafish embryos to study the general toxicity and cardiovascular effects of eight methylxanthines: aminophylline, caffeine, diprophylline, doxofylline, etophylline, 3-isobutyl-1-methylxanthine (IBMX), pentoxifylline and theophylline. Microinjections of the eight methylxanthines were performed in 1-2 cell stage zebrafish embryos and the general toxicity and cardiovascular effects were analyzed at different time points. Embryotoxicity and teratogenicity were evaluated to understand the general toxicity of these compounds. Structural and functional alterations of the heart were evaluated to assess the cardiovascular effects. Our results showed different activity patterns of the methylxanthines drugs. Caffeine, IBMX, pentoxifylline and theophylline were highly embryotoxic and teratogenic; aminophylline, doxofylline and etophylline were embryotoxic and teratogenic only at higher doses, and diprophylline showed a minimal (<10%) embryotoxicity and teratogenicity. Most of these drugs induced structural alteration of the heart in 20-40% of the injected embryos with the maximum dose. This structural alteration was fatal with the embryos ultimately dying within 120 hpf. All the drugs induced a transient increase in heart rate at 48 hpf which returned to baseline within 96 hpf. This functional effect of methylxanthines showed similarity to the studies done in humans and other vertebrates. Our results indicate the potential toxicity and teratogenicity of different methylxanthines in the embryos during embryonic development, the most sensitive period of life. Although interspecies differences need to be considered before drawing any conclusion, our study elucidated that a single exposure of methylxanthines at therapeutic range could induce cardiac dysfunction besides causing embryotoxicity and teratogenicity. Of all the drugs, diprophylline appeared to be safer, with lower degree of embryotoxicity, teratogenicity and cardiac toxicity as compared to other methylxanthines.
Sigma-Aldrich
Aminophylline, ≥98%, powder