Merck
  • RANK rewires energy homeostasis in lung cancer cells and drives primary lung cancer.
RANK rewires energy homeostasis in lung cancer cells and drives primary lung cancer.
Genes & development (2017-11-10)
Shuan Rao, Verena Sigl, Reiner Alois Wimmer, Maria Novatchkova, Alexander Jais, Gabriel Wagner, Stephan Handschuh, Iris Uribesalgo, Astrid Hagelkruys, Ivona Kozieradzki, Luigi Tortola, Roberto Nitsch, Shane J Cronin, Michael Orthofer, Daniel Branstetter, Jude Canon, John Rossi, Manolo D'Arcangelo, Johan Botling, Patrick Micke, Linnea La Fleur, Karolina Edlund, Michael Bergqvist, Simon Ekman, Thomas Lendl, Helmut Popper, Hiroshi Takayanagi, Lukas Kenner, Fred R Hirsch, William Dougall, Josef M Penninger
ABSTRACT
Lung cancer is the leading cause of cancer deaths. Besides smoking, epidemiological studies have linked female sex hormones to lung cancer in women; however, the underlying mechanisms remain unclear. Here we report that the receptor activator of nuclear factor-kB (RANK), the key regulator of osteoclastogenesis, is frequently expressed in primary lung tumors, an active RANK pathway correlates with decreased survival, and pharmacologic RANK inhibition reduces tumor growth in patient-derived lung cancer xenografts. Clonal genetic inactivation of
Sigma-Aldrich
Anti-β-Actin–FITC antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.