Merck
  • The synergistic effects of valproic acid and fluvastatin on apoptosis induction in glioblastoma multiforme cell lines.
The synergistic effects of valproic acid and fluvastatin on apoptosis induction in glioblastoma multiforme cell lines.
The international journal of biochemistry & cell biology (2017-10-12)
Yung-Lung Chang, Li-Chun Huang, Ying-Chuan Chen, Yi-Wen Wang, Dueng-Yuan Hueng, Shih-Ming Huang
ABSTRACT
Glioblastoma multiforme (GBM) is the most common primary central nervous system malignant tumor. It responds poorly to standard therapies, such as surgical resection, radiation therapy and chemotherapy. Many chemotherapeutic drugs are focused on apoptosis induction and radiation sensitivity. Inhibition of histone acetylation via histone deacetylase inhibitor (HDACI) is one such strategy. Statins (or 3-hydroxy-3-methylglutaryl-CoA reductase inhibitors) are classical drugs used to lower cholesterol but also inhibitors of histone deacetylation activity. This study analyzes the combinatory effects of valproic acid (VPA) and fluvastatin on apoptosis induction in GBM8401 cells. The results show that they act synergistically in inducing γ-H2AX and apoptosis accompanied by higher acetylated histones H3 and H4. Downregulation of p53 occurred by VPA alone and fluvastatin alone, but not at their combined application; upregulation of p21 at the protein level was induced by each of the drugs alone and no further increase occurred at combined application. The drug BEZ235 inhibited phosphorylation of Akt and attenuated the level of γ-H2AX as well as cleaved PARP (cPARP) induced at combined application of VPA and fluvastatin. Induction of apoptosis within a 48h incubation period was massive when measured as the subG1 peak (97%) and was detected after a 24h incubation at low level when assayed with PE Annexin V. Synergistic apoptosis induction was demonstrated also after 24h incubation by the appearance of cPARP. Partial silencing of p21 reduced cPARP as well as the percentage of apoptotic cells in the subG1 peak. However, partial silencing of p53 had no effect on apoptosis. Such findings offer a better understanding of the mechanism of action of HDACIs in combination with statins that may guide the development of a new combinatory reposition for the treatment of GBM.
Sigma-Aldrich
SAHA, ≥98% (HPLC)