Merck
  • Novel ATP-competitive Akt inhibitor afuresertib suppresses the proliferation of malignant pleural mesothelioma cells.
Novel ATP-competitive Akt inhibitor afuresertib suppresses the proliferation of malignant pleural mesothelioma cells.
Cancer medicine (2017-09-30)
Masayuki Yamaji, Akinobu Ota, Md Wahiduzzaman, Sivasundaram Karnan, Toshinori Hyodo, Hiroyuki Konishi, Shinobu Tsuzuki, Yoshitaka Hosokawa, Masayuki Haniuda
ABSTRACT
Malignant pleural mesothelioma (MPM), an asbestos-related occupational disease, is an aggressive and incurable tumor of the thoracic cavity. Despite recent advances in MPM treatment, overall survival of patients with MPM is very low. Recent studies have implicated that PI3K/Akt signaling is involved in MPM cell survival and development. To investigate the effects of Akt inhibitors on MPM cell survival, we examined the effects of nine selective Akt inhibitors, namely, afuresertib, Akti-1/2, AZD5363, GSK690693, ipatasertib, MK-2206, perifosine, PHT-427, and TIC10, on six MPM cell lines, namely, ACC-MESO-4, Y-MESO-8A, MSTO-211H, NCI-H28, NCI-H290, and NCI-H2052, and a normal mesothelial cell line MeT-5A. Comparison of IC
Sigma-Aldrich
GSK690693, ≥98% (HPLC)