Merck
  • Identification of a New Morpholine Scaffold as a P2Y12 Receptor Antagonist.
Identification of a New Morpholine Scaffold as a P2Y12 Receptor Antagonist.
Molecules (Basel, Switzerland) (2016-08-27)
Young Ha Ahn, Joo-Youn Lee, Hee Dong Park, Tae Hun Kim, Min Chul Park, Gildon Choi, Sunghoon Kim
ABSTRACT
The P2Y12 receptor is critical for platelet activation and is an attractive drug target for the prevention of atherothrombotic events. Despite the proven antithrombotic efficacy of P2Y12 inhibitors, these thienopyridine scaffolds are prodrugs that lack important features of the ideal antithrombotic agent. For this reason, ticagrelor-a new chemical class of P2Y12 receptor antagonist-was developed, but it can cause shortness of breath and various types of bleeding. Moreover, ticagrelor is a cytochrome P450 3A4 substrate/inhibitor and, therefore, caution should be exercised when it is used concomitantly with strong CYP3A4 inducers/inhibitors. There is a need for novel P2Y12 receptor antagonist scaffolds that are reversible and have high efficacy without associated side effects. Here, we describe a novel antagonist containing a morpholine moiety that was identified by screening libraries of commercially available compounds. The molecule, Compound E, acted on P2Y12, but not P2Y1 and P2Y13, and exhibited pharmacological characteristics that were distinct from those of ticagrelor, acting instead on P2Y12 via an allosteric mechanism. These results provide a basis for the development/optimization of a new class of P2Y12 antagonists.
Sigma-Aldrich
AZD1283, ≥98% (HPLC)