Merck
  • Spatial detection of fetal marker genes expressed at low level in adult human heart tissue.
Spatial detection of fetal marker genes expressed at low level in adult human heart tissue.
Scientific reports (2017-10-13)
Michaela Asp, Fredrik Salmén, Patrik L Ståhl, Sanja Vickovic, Ulrika Felldin, Marie Löfling, José Fernandez Navarro, Jonas Maaskola, Maria J Eriksson, Bengt Persson, Matthias Corbascio, Hans Persson, Cecilia Linde, Joakim Lundeberg
ABSTRACT
Heart failure is a major health problem linked to poor quality of life and high mortality rates. Hence, novel biomarkers, such as fetal marker genes with low expression levels, could potentially differentiate disease states in order to improve therapy. In many studies on heart failure, cardiac biopsies have been analyzed as uniform pieces of tissue with bulk techniques, but this homogenization approach can mask medically relevant phenotypes occurring only in isolated parts of the tissue. This study examines such spatial variations within and between regions of cardiac biopsies. In contrast to standard RNA sequencing, this approach provides a spatially resolved transcriptome- and tissue-wide perspective of the adult human heart, and enables detection of fetal marker genes expressed by minor subpopulations of cells within the tissue. Analysis of patients with heart failure, with preserved ejection fraction, demonstrated spatially divergent expression of fetal genes in cardiac biopsies.
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Sigma-Aldrich
SSC Buffer 20× Concentrate, for Northern and Southern blotting, solution