Merck
  • Design, synthesis and evaluation of coumarin-pargyline hybrids as novel dual inhibitors of monoamine oxidases and amyloid-β aggregation for the treatment of Alzheimer's disease.
Design, synthesis and evaluation of coumarin-pargyline hybrids as novel dual inhibitors of monoamine oxidases and amyloid-β aggregation for the treatment of Alzheimer's disease.
European journal of medicinal chemistry (2017-07-21)
Hua-Li Yang, Pei Cai, Qiao-Hong Liu, Xue-Lian Yang, Fan Li, Jin Wang, Jia-Jia Wu, Xiao-Bing Wang, Ling-Yi Kong
ABSTRACT
A series of coumarin-pargyline hybrids (4a-x) have been designed, synthesized and evaluated as novel dual inhibitors of Alzheimer's disease (AD). Most of the compounds exhibited a potent ability to inhibit amyloid-β (Aβ) aggregation and monoamine oxidases. In particular, compound 4x exhibited remarkable inhibitory activities against monoamine oxidases (IC
Sigma-Aldrich
3-Fluorobenzylamine, 97%