Merck
  • Lin28 is induced in primed embryonic stem cells and regulates let-7-independent events.
Lin28 is induced in primed embryonic stem cells and regulates let-7-independent events.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-12-07)
Silvia Parisi, Fabiana Passaro, Luigi Russo, Anna Musto, Angelica Navarra, Simona Romano, Giuseppe Petrosino, Tommaso Russo
ABSTRACT
Lin28 RNA-binding proteins play important roles in pluripotent stem cells, but the regulation of their expression and the mechanisms underlying their functions are still not definitively understood. Here we address the above-mentioned issues in the first steps of mouse embryonic stem cell (ESC) differentiation. We observed that the expression of
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hmga2
Sigma-Aldrich
Triton X-100, laboratory grade