Merck
  • 'Candidatus Dichloromethanomonas elyunquensis' gen. nov., sp. nov., a dichloromethane-degrading anaerobe of the Peptococcaceae family.
'Candidatus Dichloromethanomonas elyunquensis' gen. nov., sp. nov., a dichloromethane-degrading anaerobe of the Peptococcaceae family.
Systematic and applied microbiology (2017-03-16)
Sara Kleindienst, Steven A Higgins, Despina Tsementzi, Gao Chen, Konstantinos T Konstantinidis, E Erin Mack, Frank E Löffler
ABSTRACT
Taxonomic assignments of anaerobic dichloromethane (DCM)-degrading bacteria remain poorly constrained but are important for understanding the microbial diversity of organisms contributing to DCM turnover in environmental systems. We describe the taxonomic classification of a novel DCM degrader in consortium RM obtained from pristine Rio Mameyes sediment. Phylogenetic analysis of full-length 16S rRNA gene sequences demonstrated that the DCM degrader was most closely related to members of the genera Dehalobacter and Syntrophobotulus, but sequence similarities did not exceed 94% and 93%, respectively. Genome-aggregate average amino acid identities against Peptococcaceae members did not exceed 66%, suggesting that the DCM degrader does not affiliate with any described genus. Phylogenetic analysis of conserved single-copy functional genes supported that the DCM degrader represents a novel clade. Growth strictly depended on the presence of DCM, which was consumed at a rate of 160±3μmolL
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%