Merck
Pharmacological properties of SAK3, a novel T-type voltage-gated Ca
Neuropharmacology (2017-01-18)
Yasushi Yabuki, Kazuya Matsuo, Hisanao Izumi, Hidaka Haga, Takashi Yoshida, Minoru Wakamori, Akikazu Kakei, Kenji Sakimura, Takaichi Fukuda, Kohji Fukunaga
ABSTRACT
T-type voltage-gated Ca
Sigma-Aldrich
NNC 55-0396 hydrate, ≥98% (HPLC)
Custom Peptides, Product Overview, Design, and Ordering
Sigma-Aldrich
SAK3, ≥98% (HPLC)