Merck
  • Crystal structures of human SIRT3 displaying substrate-induced conformational changes.
Crystal structures of human SIRT3 displaying substrate-induced conformational changes.
The Journal of biological chemistry (2009-06-19)
Lei Jin, Wentao Wei, Yaobin Jiang, Hao Peng, Jianhua Cai, Chen Mao, Han Dai, Wendy Choy, Jean E Bemis, Michael R Jirousek, Jill C Milne, Christoph H Westphal, Robert B Perni
ABSTRACT
SIRT3 is a major mitochondrial NAD(+)-dependent protein deacetylase playing important roles in regulating mitochondrial metabolism and energy production and has been linked to the beneficial effects of exercise and caloric restriction. SIRT3 is emerging as a potential therapeutic target to treat metabolic and neurological diseases. We report the first sets of crystal structures of human SIRT3, an apo-structure with no substrate, a structure with a peptide containing acetyl lysine of its natural substrate acetyl-CoA synthetase 2, a reaction intermediate structure trapped by a thioacetyl peptide, and a structure with the dethioacetylated peptide bound. These structures provide insights into the conformational changes induced by the two substrates required for the reaction, the acetylated substrate peptide and NAD(+). In addition, the binding study by isothermal titration calorimetry suggests that the acetylated peptide is the first substrate to bind to SIRT3, before NAD(+). These structures and biophysical studies provide key insight into the structural and functional relationship of the SIRT3 deacetylation activity.