Merck
  • Disrupting ceramide-CD300f interaction prevents septic peritonitis by stimulating neutrophil recruitment.
Disrupting ceramide-CD300f interaction prevents septic peritonitis by stimulating neutrophil recruitment.
Scientific reports (2017-06-29)
Kumi Izawa, Akie Maehara, Masamichi Isobe, Yuka Yasuda, Makoto Urai, Yasutaka Hoshino, Keigo Ueno, Toshihiro Matsukawa, Mariko Takahashi, Ayako Kaitani, Emiko Shiba, Ayako Takamori, Shino Uchida, Koichiro Uchida, Keiko Maeda, Nobuhiro Nakano, Yoshinori Yamanishi, Toshihiko Oki, David Voehringer, Axel Roers, Susumu Nakae, Junko Ishikawa, Yuki Kinjo, Toshiaki Shimizu, Hideoki Ogawa, Ko Okumura, Toshio Kitamura, Jiro Kitaura
ABSTRACT
Sepsis is a serious clinical problem. Negative regulation of innate immunity is associated with sepsis progression, but the underlying mechanisms remains unclear. Here we show that the receptor CD300f promotes disease progression in sepsis. CD300f
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)