Merck
Efficient Generation of Lactic Acid from Glycerol over a Ru-Zn-Cu
Chemistry, an Asian journal (2017-05-04)
Zhiwei Jiang, Zhanrong Zhang, Tianbin Wu, Pei Zhang, Jinliang Song, Chao Xie, Buxing Han
ABSTRACT
The biodiesel production process generates a significant amount of glycerol as a byproduct. A drive to add value has attracted worldwide attention, with the aim of improving the overall effectiveness and profitability of biodiesel production. Herein, we report hydroxyapatite (HAP)-supported Ru-Zn-Cu
Sigma-Aldrich
Ruthenium(III) chloride trihydrate, technical