Merck
Network model predicts that CatSper is the main Ca
Scientific reports (2017-06-28)
Jesús Espinal-Enríquez, Daniel Alejandro Priego-Espinosa, Alberto Darszon, Carmen Beltrán, Gustavo Martínez-Mekler
ABSTRACT
Spermatozoa sea urchin swimming behaviour is regulated by small peptides from the egg outer envelope. Speract, such a peptide, after binding to its receptor in Strongylocentrotus purpuratus sperm flagella, triggers a signaling pathway that culminates with a train of intracellular calcium oscillations, correlated with changes in sperm swimming pattern. This pathway has been widely studied but not fully characterized. Recent work on Arbacia punctulata sea urchin spermatozoa has documented the presence of the Ca
Sigma-Aldrich
NNC 55-0396 hydrate, ≥98% (HPLC)