- Seed Pelleting with Calcium Peroxide Improves Crop Establishment of Direct-seeded Rice under Waterlogging Conditions.
Seed Pelleting with Calcium Peroxide Improves Crop Establishment of Direct-seeded Rice under Waterlogging Conditions.
Scientific reports (2017-07-09)
Junhao Mei, Weiqin Wang, Shaobing Peng, Lixiao Nie
PMID28687785
ABSTRACT
Poor crop establishment of direct-seeded rice (DSR) is one of the major constraints to wide adoption of DSR, particularly in areas prone to flooding after sowing or where fields are not level. Seed pelleting is an effective, practical and facile technique to enhance crop establishment under unfavorable environmental conditions. To evaluate the effects of seed pelleting on rice germination, seedling growth and associated metabolic events under waterlogging stress, various seed pelleting treatments including formulae, pelleting times (the weight ratio of pelleting agents: rice seeds = 1:1~7:1 (w/w) and CaO