Merck
  • Down-regulation of DLC1 in endothelial cells compromises the angiogenesis process.
Down-regulation of DLC1 in endothelial cells compromises the angiogenesis process.
Cancer letters (2017-04-15)
Yi-Ping Shih, Sarah Y Yuan, Su Hao Lo
ABSTRACT
DLC1 is a RhoGAP-containing tumor suppressor that inhibits angiogenesis by repressing VEGF production in epithelial cells. Here we report the roles of DLC1 in endothelial cells. Silencing of DLC1 (siDLC1) enhances cell migration but reduces tube formation activities of human umbilical vein endothelial cells (HUVECs). Biochemically, RhoA activity and paxillin protein level are markedly increased in siDLC1 HUVECs. Although further silencing of RhoA restores the cell migration phenotype, the tube formation defect and up-regulated paxillin level remain unchanged. On the other hand, paxillin knockdown rescues tube formation and migration phenotypes but not the up-regulated RhoA activity. These results indicate that DLC1 regulates endothelial cell migration through RhoA and paxillin independently and controls tube formation mainly via paxillin. To further determine endothelial DLC1's function, we have generated endothelial specific knockout mice (DLC1-Tek). DLC1-Tek mice appear to be normal and healthy but their angiogenesis processes are compromised as shown in gel plug and aortic ring sprouting assays. Analysis of endothelial cells isolated from DLC1-Tek mice has further affirmed the cellular and biochemical phenotypes established in siDLC1 HUVECs. Our studies have demonstrated a positive regulatory role of endothelial DLC1 in angiogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human PXN (1)