Merck
  • Kindlin-2 expression in peritumoral stroma is associated with poor prognosis in pancreatic ductal adenocarcinoma.
Kindlin-2 expression in peritumoral stroma is associated with poor prognosis in pancreatic ductal adenocarcinoma.
Pancreas (2013-03-20)
Prawej Mahawithitwong, Kenoki Ohuchida, Naoki Ikenaga, Hayato Fujita, Ming Zhao, Shingo Kozono, Koji Shindo, Takao Ohtsuka, Kazuhiro Mizumoto, Masao Tanaka
ABSTRACT
Kindlin-2 is a novel focal adhesion protein reported to be expressed in breast, lung, and gastric cancers. This study aimed to investigate the significance of kindlin-2 expression in pancreatic ductal adenocarcinomas (PDACs). We performed immunohistochemical analysis on kindlin-2 on PDAC samples from 95 patients. We investigated the association between kindlin-2 expression and clinicopathological parameters of PDAC and the survival time of patients with PDAC who underwent pancreatectomy. Kindlin-2 was highly expressed in the peritumoral stroma of PDACs. Stromal kindlin-2 expression was related to nodal metastasis (P = 0.03). Univariate analysis showed that patients with positive kindlin-2 expression had significantly shorter survival times than those with negative kindlin-2 expression (P = 0.01). In addition, multivariate analysis revealed that kindlin-2 expression was an independent factor of poor prognosis in patients with PDAC after R0 resection (RR = 2.15; P = 0.04). Kindlin-2 expression in stromal components is significantly associated with poor prognosis of patients with PDAC, suggesting that kindlin-2 is a prognostic marker for patients with PDAC.