Merck
  • Radical anion and dianion salts of titanyl macrocycles with acceptor substituents or an extended π-system.
Radical anion and dianion salts of titanyl macrocycles with acceptor substituents or an extended π-system.
Dalton transactions (Cambridge, England : 2003) (2017-02-28)
Maxim A Faraonov, Dmitri V Konarev, Alexey M Fatalov, Salavat S Khasanov, Sergey I Troyanov, Rimma N Lyubovskaya
ABSTRACT
Crystalline anionic salts of titanyl macrocycles with acceptor substituents or an extended π-system have been obtained for the first time: (PPN
Sigma-Aldrich
Diaminomaleonitrile, 98%
Sigma-Aldrich
Acenaphthenequinone
Sigma-Aldrich
4,5-Dichlorophthalonitrile, 99%