Merck
  • A biocompatible macromolecular two-photon initiator based on hyaluronan.
A biocompatible macromolecular two-photon initiator based on hyaluronan.
Polymer chemistry (2017-03-07)
Maximilian Tromayer, Peter Gruber, Marica Markovic, Arnulf Rosspeintner, Eric Vauthey, Heinz Redl, Aleksandr Ovsianikov, Robert Liska
ABSTRACT
The possibility of the direct encapsulation of living cells
Sigma-Aldrich
tert-Butyl carbamate, 98%