Merck
  • A point mutation in the tyrosine hydroxylase gene associated with Segawa's syndrome.
A point mutation in the tyrosine hydroxylase gene associated with Segawa's syndrome.
Human genetics (1995-01-01)
B Lüdecke, B Dworniczak, K Bartholomé
ABSTRACT
We have examined the molecular basis of Segawa's syndrome in six families with seven affected children. In one family two siblings with this disease carried a point mutation in exon 11 of the tyrosine hydroxylase gene, resulting in an amino acid exchange of Gln381 to Lys381. These results suggest that a change in tyrosine hydroxylase causes this form of Segawa's syndrome.