Merck
  • A cyclopropanol-based strategy for subunit coupling: total synthesis of (+)-spirolaxine methyl ether.
A cyclopropanol-based strategy for subunit coupling: total synthesis of (+)-spirolaxine methyl ether.
Organic letters (2007-06-15)
Katie A Keaton, Andrew J Phillips
ABSTRACT
A strategy for ketone synthesis with cyclopropanols as intermediates and its application to (+)-spirolaxine methyl ether is described. The synthesis also features an application of Fu's alkyl-alkyl Suzuki coupling.
Sigma-Aldrich
3,5-Dimethoxybenzaldehyde, 98%