Merck
Cyclizations of phenylethyl-substituted pyridinecarboxaldehydes.
Tetrahedron (2013-10-22)
Rajasekhar Reddy Naredla, Douglas A Klumpp
ABSTRACT
Several phenylethyl-substituted pyridinecarboxaldehydes were prepared from 2-bromo-3-pyridinecarboxaldehyde and these substances are found to undergo cyclization reactions in acidic media. In the absence of added nucleophile, acid promoted cyclization and oxidation (MnO