Merck
Ionic bis-nanoparticle networks.
Monatshefte fur chemie (2012-01-01)
Marie-Alexandra Neouze, Marco Litschauer, Michael Puchberger, Johannes Bernardi
ABSTRACT
A newly arising challenge in the field of nanoparticle research concerns the control and the understanding of the interparticle interactions and interparticle properties. This should allow the development of materials based on nanoparticle assemblies which represents a great opportunity to exploit nanoparticle collective properties. Although some nanoparticle networks have been reported, few works are addressing the highly exciting problem of forming bis-nanoparticle assemblies in which two different types of nanoparticles are present. In this article we report an original synthesis pathway for the formation of an ionic bis-nanoparticle network, silica/silver, based on the formation of an imidazolium bridging unit. The reaction used for the formation of the bridging imidazolium can be considered as click-like chemistry. The synthesis of the metal/metal oxide hybrid composite material starts from the formation of a metal oxide nanoparticle modified with an imidazole ligand. This composite formation is therefore very general and could be extended to other metal/metal oxide composites. .
Sigma-Aldrich
(3-Chloropropyl)trimethoxysilane, ≥97%