Merck
  • Characterization of the endokinins: human tachykinins with cardiovascular activity.
Characterization of the endokinins: human tachykinins with cardiovascular activity.
Proceedings of the National Academy of Sciences of the United States of America (2003-04-30)
Nigel M Page, Nicola J Bell, Sheila M Gardiner, Isaac T Manyonda, Kerensa J Brayley, Philip G Strange, Philip J Lowry
ABSTRACT
We report four human tachykinins, endokinins A, B, C, and D (EKA-D), encoded from a single tachykinin precursor 4 gene that generates four mRNAs (alpha, beta, gamma, and delta). Tachykinin 4 gene expression was detected primarily in adrenal gland and in the placenta, where, like neurokinin B, significant amounts of EKB-like immunoreactivity were detected. EKA/B 10-mers displayed equivalent affinity for the three tachykinin receptors as substance P (SP), whereas a 32-mer N-terminal extended form of EKB was significantly more potent than EKA/B or SP. EKC/D, which possess a previously uncharacterized tachykinin motif, FQGLL-NH(2), displayed low potency. EKA/B displayed identical hemodynamic effects to SP in rats, causing short-lived falls in mean arterial blood pressure associated with tachycardia, mesenteric vasoconstriction, and marked hindquarter vasodilatation. Thus, EKA/B could be the endocrineparacrine agonists at peripheral SP receptors and there may be as yet an unidentified receptor(s) for EKC/D.
Sigma-Aldrich
Hemokinin 1 human, ≥98% (HPLC), solid