Merck
  • P(i-BuNCH2CH2)3N: an efficient ligand for the direct alpha-arylation of nitriles with aryl bromides.
P(i-BuNCH2CH2)3N: an efficient ligand for the direct alpha-arylation of nitriles with aryl bromides.
The Journal of organic chemistry (2003-10-11)
Jingsong You, John G Verkade
ABSTRACT
A new catalyst system for the synthesis of alpha-aryl-substituted nitriles is reported. The bicyclic triaminophosphine P(i-BuNCH(2)CH(2))(3)N (1b) serves as an efficient and versatile ligand for the palladium-catalyzed direct alpha-arylation of nitriles with aryl bromides. Using ligand 1b, ethyl cyanoacetate and primary as well as secondary nitriles are efficiently coupled with a wide variety of aryl bromides possessing electron-rich, electron-poor, electron-neutral, and sterically hindered groups.
Sigma-Aldrich
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, 97%