Merck
  • Expression of interferon-gamma in human adrenal gland and kidney tumours.
Expression of interferon-gamma in human adrenal gland and kidney tumours.
British journal of cancer (2007-07-12)
Q Li, X-Q Zhang, L Nie, G-S Chen, H Li, F Zhang, L-Y Zhang, L Hong, S-F Wang, H Wang
ABSTRACT
It is known that interferon-gamma (IFN-gamma) is produced by activated T and NK lymphoid cells, mononuclear cells, and macrophage and dendritic cells. Our previous studies have shown that IFN-gamma-like immunoreactivity also appears in human adrenal cortical tumour and phaeochromocytoma. To investigate whether human tumour cells can produce IFN-gamma, we examined 429 biopsy specimens of 30 kinds of tumour and tumour-surrounding tissues in adrenal glands and in kidneys by using immunohistochemistry and in situ hybridisation. IFN-gamma immunoactivity was shown in 34.3% of the adrenal cortical adenomas, 50% of the adrenal cortical carcinomas, 26.7% of the phaeochromocytomas, 26.7% of the clear cell renal cell carcinomas (RCCs), 22% of the adrenal cortexes and 40% of medullas adjacent to tumours. The positive samples and expression areas were well overlapped between the IFN-gamma mRNA and the immunohistochemistry staining. Western blot analysis has further confirmed the immunohistochemistry results by showing a distinct IFN-gamma band corresponding to 17.4 kDa in tissue extracts from adrenal cortical adenoma, phaeochromocytoma and clear cell RCCs. These results indicate that IFN-gamma is produced by some types of tumour cells, suggesting it may play a dual role in the development of these tumours.
Product Description
Supelco
Color Marker Ultra-low Range (M.W. 1,060-26,600)