Merck
  • Synthesis of D-glucose 3- and 6-[2-(perfluoroalkyl)ethyl] phosphates: a new type of anionic surfactant for biomedical use.
Synthesis of D-glucose 3- and 6-[2-(perfluoroalkyl)ethyl] phosphates: a new type of anionic surfactant for biomedical use.
Carbohydrate research (1992-05-22)
A Milius, J Greiner, J G Riess
ABSTRACT
D-Glucose 3- and 6-[sodium 2-(perfluoro-hexyl or -octyl)ethyl phosphates) have been synthesized by condensation of 1,2,3,4,-tetra-O-acetyl-beta-D-glucopyranose and 1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose with 2-(perfluoroalkyl)ethylphosphoroditriazolides followed by O-deacetylation or deacetalation. The structures of the compounds were established on the basis of 1H-, 19F-, 31P-, and 13C-NMR data. These salts display strong surface activities and appear to have good biocompatibility.
Sigma-Aldrich
1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose, 98%