Merck
  • Simple preparation of Pd-NP/polythiophene nanospheres for heterogeneous catalysis.
Simple preparation of Pd-NP/polythiophene nanospheres for heterogeneous catalysis.
Journal of colloid and interface science (2015-06-24)
Sang-Eun Bae, Ki-Jung Kim, Yong-Kyung Hwang, Seong Huh
ABSTRACT
A very simple preparation was developed for catalytically active Pd-nanoparticles (Pd-NPs) decorating polythiophene conducting polymer nanospheres by the redox reaction between PdCl4(2-) ion and 2-thiophenemethanol (2-TPM) in an aqueous solution at room temperature. 2-TPM polymerized to form polythiophene nanospheres in the presence of PdCl4(2-) ions, reduced to Pd-NPs without the need for extra reducing agents or organic surface capping ligands for sub-20 nm Pd-NPs that uniformly cover polythiophene nanospheres whose dimensions range from 120 nm to 200 nm. The Pd-NP/polythiophene nanospheres were characterized by scanning transmission electron microscopy (STEM), transmission electron microscopy (TEM), and scanning electron microscopy (SEM), powder X-ray diffraction (PXRD), X-ray photoelectron spectroscopy (XPS), and inductively-coupled plasma atomic emission spectroscopy (ICP-AES). The Pd-NP/polythiophene nanospheres were found to be an excellent catalyst for Suzuki-Miyaura cross-coupling reaction for a wide range of substrates under mild aerobic reaction conditions.
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Thiophene, ≥99%