Merck
  • CCR4 and CXCR3 play different roles in the migration of T cells to inflammation in skin, arthritic joints, and lymph nodes.
CCR4 and CXCR3 play different roles in the migration of T cells to inflammation in skin, arthritic joints, and lymph nodes.
European journal of immunology (2014-04-05)
Nadia A Al-Banna, Maria Vaci, Drew Slauenwhite, Brent Johnston, Thomas B Issekutz
ABSTRACT
CCR4 and CXCR3 are expressed on several T-cell subsets in inflamed tissues, yet their role in tissue-specific recruitment is unclear. We examined the contributions of CCR4 and CXCR3 to T-cell recruitment into inflamed joints in collagen-induced arthritis, antigen-draining lymph nodes (LNs) and dermal inflammatory sites (poly I:C, LPS, concanavalin A, and delayed type hypersensitivity), using labeled activated T cells from CXCR3(-/-), CCR4(-/-), and WT mice. Both CXCR3 and CCR4 deficiency reduced the development of arthritis, but did not affect Th1-cell recruitment to the inflamed joints. Accumulation in inflamed LNs was highly CXCR3 dependent. In contrast, CCR4-deficient Th1 cells had an increased accumulation in these LNs. Migration to all four dermal inflammatory sites by activated Th1 and T cytotoxic cells and memory CD4(+) T cells was partially CXCR3-dependent, but Treg-cell migration was independent of CXCR3. The subset of cells expressing CCR4 has skin-migrating properties, but CCR4 itself is not required for the migration. Thus, migration into these inflamed tissues is CCR4-independent, and partially dependent on CXCR3, except for Treg cells, which require neither receptor. CCR4 may therefore affect retention of T cells in different tissues rather than trafficking out of the blood.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
8-Quinolinol, PESTANAL®, analytical standard
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
8-Hydroxyquinoline
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 98.5%
Supelco
Rapamycin, VETRANAL®, analytical standard