Merck
  • Phosphodiesterase 4 interacts with the 5-HT4(b) receptor to regulate cAMP signaling.
Phosphodiesterase 4 interacts with the 5-HT4(b) receptor to regulate cAMP signaling.
Cellular signalling (2014-08-08)
S Weninger, K Van Craenenbroeck, R T Cameron, F Vandeput, M A Movsesian, G S Baillie, R A Lefebvre
ABSTRACT
Phosphodiesterase (PDE) 3 and PDE4, which degrade cyclic adenosine monophosphate (cAMP), are important regulators of 5-hydroxytryptamine (5-HT) 4 receptor signaling in cardiac tissue. Therefore, we investigated whether they interact with the 5-HT4(b) receptor, and whether A-kinase anchoring proteins (AKAPs), scaffolding proteins that bind to the regulatory subunit of protein kinase A (PKA) and contribute to the spacial-temporal control of cAMP signaling, are involved in the regulation of 5-HT4(b) receptor signaling. By measuring PKA activity in the absence and presence of PDE3 and PDE4 inhibitiors, we found that constitutive signaling of the overexpressed HA-tagged 5-HT4(b) receptor in HEK293 cells is regulated predominantly by PDE4, with a secondary role for PDE3 that is unmasked in the presence of PDE4 inhibition. Overexpressed PDE4D3 and PDE3A1, and to a smaller extent PDE4D5 co-immunoprecipitate constitutively with the 5-HT4(b) receptor. PDE activity measurements in immunoprecipitates of the 5-HT4(b) receptor confirm the association of PDE4D3 with the receptor and provide evidence that the activity of this PDE may be increased upon receptor stimulation with 5-HT. A possible involvement of AKAPs in 5-HT4(b) receptor signaling was uncovered in experiments using the St-Ht31 inhibitor peptide, which disrupts the interaction of AKAPs with PKA. However, St-Ht31 did not influence 5-HT4(b) receptor-stimulated PKA activity, and endogenous AKAP79 and gravin were not found in immunoprecipitates of the 5-HT4(b) receptor. In conclusion, we found that both PDE3A1 and PDE4D3 are integrated into complexes that contain the 5-HT4(b) receptor and may thereby regulate 5-HT4(b) receptor-mediated signaling.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Isoprenaline hydrochloride
Sigma-Aldrich
Cilostamide, phosphodiesterase inhibitor
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Isoprenaline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)