Merck
  • The neurotensin-1 receptor agonist PD149163 inhibits conditioned avoidance responding without producing catalepsy in rats.
The neurotensin-1 receptor agonist PD149163 inhibits conditioned avoidance responding without producing catalepsy in rats.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology (2011-02-01)
Elizabeth N Holly, Bree Ebrecht, Adam J Prus
ABSTRACT
Agonists for neurotensin (NT)-1 receptors have produced antipsychotic-like effects in many animals, including reversal of prepulse inhibition deficits and psychostimulant-induced increases in spontaneous activity. The present study sought to provide a basic assessment of the putative antipsychotic effects of PD149163 in rats using a two way conditioned avoidance response task, which is highly validated for screening antipsychotic drugs, and an inclined grid assessment, which is used to assess extrapyramidal side effect liability. PD149163 (0.0625-8.0 mg/kg) significantly suppressed conditioned avoidance responding (CAR) following administration of a 1.0 or 8.0 mg/kg dose. PD149163 failed to significantly increase catalepsy scores. The typical antipsychotic drug haloperidol (0.01-1.0 mg/kg) significantly suppressed CAR at a 0.1, 0.3, and 1.0 mg/kg dose, and a significant increase in catalepsy scores was found at the 1.0 mg/kg dose. The atypical antipsychotic drug clozapine (2.5-10.0 mg/kg) also produced a significant inhibition of CAR, which occurred following administration of a 10.0 mg/kg dose. Clozapine failed to significantly increase catalepsy scores. Finally, D-amphetamine (1.0 mg/kg), serving as a negative control, failed to suppress CAR or increase catalepsy scores. These data further suggest that PD149163 may have atypical antipsychotic-like properties.
Sigma-Aldrich
PD 149163 tetrahydrochloride hydrate, ≥90% (HPLC)