Merck
  • Monitoring the coordination of amine ligands on silver nanoparticles using NMR and SERS.
Monitoring the coordination of amine ligands on silver nanoparticles using NMR and SERS.
Langmuir : the ACS journal of surfaces and colloids (2015-01-08)
Jérémy Cure, Yannick Coppel, Thameur Dammak, Pier Francesco Fazzini, Adnen Mlayah, Bruno Chaudret, Pierre Fau
ABSTRACT
Low size dispersity silver nanoparticles (ca. 6 nm) have been synthesized by the hydrogenolysis of silver amidinate in the presence of hexadecylamine. Combining NMR techniques with SERS and DFT modeling, it is possible to observe an original stabilization mechanism. Amidine moiety is strongly coordinated to the Ag(0) nanoparticles surface whereas HDA ligand is necessary to prevent agglomeration, although it is only weakly interacting with the surface.
Sigma-Aldrich
Hexadecylamine, 98%